Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027413.1 | NP_081689.1 | 2 | 210 | ||
NM_175124.5 | NP_780333.3 | 2 | 210 | |||
GenBank mRNA | AI663251.1 | - | 2 | 210 | ||
AK004904.1 | - | 2 | 179 | |||
AK008713.1 | - | 2 | 178 | |||
AK050154.1 | - | 2 | 182 | |||
AK077270.1 | - | 2 | 206 | |||
AK159417.1 | - | 2 | 179 | |||
AK183719.1 | - | 1 | 101 | |||
BC052051.1 | - | 1 | 117 |