Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppip5k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173760.5 | NP_776121.4 | 28 | 4067 | ||
GenBank mRNA | AK052286.1 | - | 7 | 864 | ||
AK087712.1 | - | 28 | 4001 | |||
AK129140.1 | - | 28 | 4003 | |||
AK137392.1 | - | 3 | 2559 | |||
BC046411.1 | - | 18 | 2380 | |||
BC053396.1 | - | 27 | 3569 | 30044788 |