Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyctk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039586.1 | NP_001034675.1 | 5 | 3840 | ||
NM_174846.4 | NP_777271.3 | 4 | 3976 | |||
GenBank mRNA | AK052709.1 | - | 4 | 3915 | ||
BB747408.1 | - | 1 | 268 | |||
BC025935.1 | - | 3939 | 5097665 | |||
BC036136.1 | - | 3799 | 4971642 |