Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdhx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175094.5 | NP_780303.1 | 1, 2 | 231 | ||
GenBank mRNA | AK046020.1 | - | 1, 2 | 180 | ||
AK047670.1 | - | 1, 2 | 229 | |||
AK050433.1 | - | 1, 2 | 232 | |||
AK083170.1 | - | 1, 2 | 195 | |||
AK166257.1 | - | 1, 2 | 194 | |||
BC061231.1 | - | 1, 2 | 192 | 6532760 |