Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2o | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173755.3 | NP_776116.2 | 18 | 4805 | ||
GenBank mRNA | AK035970.1 | - | 1 | 2024 | ||
AK045398.1 | - | 5 | 2175 | |||
AK129431.1 | - | 18 | 4806 | |||
BC023329.1 | - | 5 | 2536 | |||
BC039233.1 | - | 10 | 3272 | |||
BC051108.1 | - | 9 | 3150 | |||
BC055067.1 | - | 4 | 1897 | |||
BC059193.1 | - | 18 | 4431 | 4160667 |