Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173762.4 | NP_776123.3 | 39, 40 | 6320 | ||
GenBank mRNA | AK049676.1 | - | 6, 7 | 1122 | ||
AY493378.1 | - | 39, 40 | 6233 | |||
BC049989.1 | - | 5, 6 | 885 | |||
BC059032.1 | - | 7, 8 | 1282 |