Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173767.3 | NP_776128.2 | 13 | 1991 | ||
GenBank mRNA | AK014452.1 | - | 12 | 1753 | ||
AK050143.1 | - | 10 | 1486 | |||
AK193251.1 | - | 1 | 203 | |||
AK195962.1 | - | 1 | 203 | |||
AK196323.1 | - | 1 | 60 | |||
AK197875.1 | - | 1 | 203 | |||
AK200147.1 | - | 1 | 202 | |||
BC026405.1 | - | 13 | 1878 | 4211657 | ||
BC089586.1 | - | 10 | 1494 | |||
BC139196.1 | - | 10 | 1379 | 8862218 | ||
BC139197.1 | - | 10 | 1379 | 8862219 |