Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubtd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173784.3 | NP_776145.2 | 3 | 938 | ||
GenBank mRNA | AK076953.1 | - | 3 | 940 | ||
AK079381.1 | - | 1 | 232 | |||
AK165107.1 | - | 277 | ||||
BC043664.1 | - | 1 | 433 | |||
BC057026.1 | - | 3 | 938 | 6825513 |