Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou6f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175006.2 | NP_778171.2 | 4 | 1130 | ||
GenBank mRNA | AK042530.1 | - | 1 | 108 | ||
AK083772.1 | - | 4 | 1130 | |||
BC094915.1 | - | 4 | 1145 | 6435959 | ||
BC138151.1 | - | 4 | 1052 | 8861170 |