Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101.3 | NP_001092.1 | 6 | 1433 | ||
GenBank mRNA | AK025375.1 | - | 6 | 1432 | ||
AK058019.1 | - | 6 | 1433 | |||
AK098751.1 | - | 7 | 1236 | |||
AK130062.1 | - | 6 | 1435 | |||
AK130157.1 | - | 6 | 1434 | |||
AK222925.1 | - | 6 | 1436 | |||
AK223032.1 | - | 6 | 1431 | |||
AK223055.1 | - | 6 | 1431 | |||
AK225414.1 | - | 6 | 1268 | |||
AK304552.1 | - | 7 | 1362 | |||
AK308277.1 | - | 6 | 1402 | |||
AK309997.1 | - | 7 | 1475 | |||
BC001301.1 | - | 6 | 1416 | 3451917 | ||
BC002409.2 | - | 6 | 1421 | 2961617 | ||
BC004251.1 | - | 6 | 1410 | 3623067 | ||
BC008633.1 | - | 5 | 1330 | |||
BC009636.1 | - | 1 | 167 | |||
BC012854.1 | - | 5 | 1304 | |||
BC013380.2 | - | 6 | 1425 | 4025526 | ||
BC013835.1 | - | 6 | 1422 | |||
BC014861.1 | - | 6 | 1423 | 3870487 | ||
BC016045.1 | - | 6 | 1424 | 4564532 | ||
V00478.1 | - | 3 | 596 | |||
X00351.1 | - | 6 | 1390 | |||
X63432.1 | - | 6 | 1421 |