Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175098.5 | NP_780307.1 | 3 | 872 | ||
GenBank mRNA | AK018135.1 | - | 3 | 873 | ||
AK159264.1 | - | 3 | 902 | |||
BC095987.1 | - | 3 | 802 | 5365989 | ||
BC117837.2 | - | 3 | 820 | 40093139 |