Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102.3 | NP_001093.1 | 21 | 3269 | ||
NM_001130004.1 | NP_001123476.1 | 22 | 3335 | |||
NM_001130005.1 | NP_001123477.1 | 21 | 3254 | |||
XM_011537265.2 | XP_011535567.1 | 21 | 3085 | |||
XM_011537266.2 | XP_011535568.1 | 21 | 3070 | |||
XM_011537267.2 | XP_011535569.1 | 21 | 3061 | |||
XM_011537268.2 | XP_011535570.1 | 21 | 3046 | |||
XM_017021720.1 | XP_016877209.1 | 21 | 3616 | |||
XM_017021721.1 | XP_016877210.1 | 21 | 3592 | |||
XM_017021722.1 | XP_016877211.1 | 21 | 3761 | |||
XM_017021723.1 | XP_016877212.1 | 21 | 3737 | |||
XM_017021725.1 | XP_016877214.1 | 21 | 3935 | |||
XM_017021726.1 | XP_016877215.1 | 21 | 3911 | |||
XM_017021727.1 | XP_016877216.1 | 21 | 3215 | |||
XM_017021728.1 | XP_016877217.1 | 21 | 3365 | |||
GenBank mRNA | AK098203.1 | - | 14 | 1988 | ||
AK124378.1 | - | 2 | 2380 | |||
AK294317.1 | - | 15 | 2457 | |||
BC015766.1 | - | 21 | 3076 | 4856734 | ||
BI494856.1 | - | 1 | 87 | |||
BX641076.1 | - | 10 | 4408 | |||
DQ496098.1 | - | 22 | 3208 | |||
X55187.1 | - | 12 | 2071 |