Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103.3 | NP_001094.1 | 4, 5 | 660 | ||
NM_001278343.1 | NP_001265272.1 | 4, 5 | 660 | |||
NM_001278344.1 | NP_001265273.1 | 4, 5 | 660 | |||
GenBank mRNA | AB209521.1 | - | 4, 5 | 605 | ||
AK294906.1 | - | 2, 3 | 171 | |||
AK297886.1 | - | 4, 5 | 613 | |||
AK315250.1 | - | 4, 5 | 660 | |||
BC047901.2 | - | 4, 5 | 592 | 6199155 | ||
BC051770.1 | - | 4, 5 | 605 | 6198688 | ||
M86406.1 | - | 4, 5 | 613 |