Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109.4 | NP_001100.3 | 17 | 1903 | ||
NM_001164489.1 | NP_001157961.1 | 17 | 1903 | |||
XM_011539117.2 | XP_011537419.2 | 18 | 2084 | |||
XM_017015465.1 | XP_016870954.1 | 18 | 2084 | |||
XM_017015466.1 | XP_016870955.1 | 18 | 2086 | |||
GenBank mRNA | AK312834.1 | - | 17 | 1868 | ||
BC064500.1 | - | 17 | 1850 | |||
BC115404.1 | - | 17 | 1822 | 40075505 | ||
D26579.1 | - | 17 | 1826 |