Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166372.1 | NP_001159844.1 | 5 | 472 | ||
NM_001166375.1 | NP_001159847.1 | 1 | 191 | |||
NM_001293784.1 | NP_001280713.1 | 6 | 681 | |||
NM_175188.4 | NP_780397.2 | 5 | 628 | |||
GenBank mRNA | AK036489.1 | - | 1 | 192 | ||
AK086052.1 | - | 6 | 640 | |||
AK089040.1 | - | 1 | 191 | |||
AK137337.1 | - | 5 | 428 | |||
AK152592.1 | - | 1 | 193 | |||
AK163081.1 | - | 1 | 192 | |||
BC066008.1 | - | 5 | 628 | 5703753 |