Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700041G16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006671.1 | - | 123 | |||
AK033542.1 | - | 685 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_175196.1 | - | 123 | Chr. - - [-] on Build |