Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175224.4 | NP_780433.1 | 1, 2 | 133 | ||
GenBank mRNA | AK077694.1 | - | 1, 2 | 134 | ||
AK152138.1 | - | 1, 2 | 152 | |||
AK153231.1 | - | 1, 2 | 152 | |||
AK153944.1 | - | 1, 2 | 141 | |||
AK165034.1 | - | 1, 2 | 150 | |||
BC096469.1 | - | 1, 2 | 124 | 3603051 |