Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175140.4 | NP_780349.3 | 4 | 642 | ||
GenBank mRNA | AB106878.1 | - | 3 | 240 | ||
AF308145.1 | - | 4 | 460 | |||
AK005217.1 | - | 4 | 384 | |||
AK038952.1 | - | 3 | 615 | |||
AK051290.1 | - | 4 | 224 | |||
BC051124.1 | - | 4 | 607 | 6528978 | ||
CF171795.1 | - | 4 | 597 |