Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sash1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175155.4 | NP_780364.3 | 6 | 632 | ||
GenBank mRNA | AJ507736.1 | - | 6 | 447 | ||
AK149872.1 | - | 6 | 584 |