Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175168.4 | NP_780377.1 | 5 | 862 | ||
GenBank mRNA | AF531873.1 | - | 5 | 712 | ||
AK053044.1 | - | 5 | 795 | |||
AK087891.1 | - | 5 | 796 | |||
AK163135.1 | - | 5 | 795 | |||
BC076578.1 | - | 5 | 862 | 30619280 |