Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175179.4 | NP_780388.2 | 2 | 384 | ||
GenBank mRNA | AK012651.1 | - | 2 | 369 | ||
AK049774.1 | - | 2 | 369 | |||
BC053442.1 | - | 2 | 355 | |||
BC096616.1 | - | 1 | 130 | |||
CK392507.1 | - | 2 | 384 |