Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175191.4 | NP_780400.2 | 3 | 2196 | ||
GenBank mRNA | AK034194.1 | - | 3 | 2150 | ||
AK048077.1 | - | 3 | 2151 | |||
AK140577.1 | - | 3 | 2136 | |||
AK141569.1 | - | 3 | 2181 | |||
BC037625.1 | - | 1 | 650 | |||
BC089509.1 | - | 3 | 2197 | 30252116 |
Set Membership: |
G-protein coupled receptor |