Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028785.3 | NP_083061.2 | 12 | 1422 | ||
GenBank mRNA | AK018051.1 | - | 11 | 1111 | ||
AK035336.1 | - | 7 | 704 | |||
AK038285.1 | - | 11 | 1170 | |||
BC141358.1 | - | 12 | 1417 | 9055970 |