Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mzt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175245.4 | NP_780454.1 | 3 | 1898 | ||
GenBank mRNA | AK082817.1 | - | 3 | 1843 | ||
AK150268.1 | - | 3 | 1874 | |||
AK151232.1 | - | 3 | 1874 | |||
AK151358.1 | - | 3 | 1874 | |||
AK151819.1 | - | 3 | 1874 | |||
AK160911.1 | - | 3 | 1883 | |||
AK168114.1 | - | 3 | 1839 | |||
AK169036.1 | - | 3 | 1848 | |||
AK170746.1 | - | 3 | 1848 | |||
BC098481.1 | - | 3 | 1859 |