Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175198.4 | NP_780407.1 | 3 | 1682 | ||
GenBank mRNA | AK029978.1 | - | 3 | 1639 | ||
AK051700.1 | - | 3 | 1683 | |||
BC119330.1 | - | 2 | 1426 | 8734079 | ||
BC119332.1 | - | 2 | 1426 | 8734081 |