Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290570.1 | NP_001277499.1 | 15, 16 | 1669 | ||
NM_001290572.1 | NP_001277501.1 | 16, 17 | 1723 | |||
NM_175211.5 | NP_780420.1 | 18, 19 | 1933 | |||
GenBank mRNA | AK076767.1 | - | 18, 19 | 1918 | ||
AK129123.1 | - | 15, 16 | 1636 | |||
AK170248.1 | - | 16, 17 | 1670 | |||
BC037725.1 | - | 4, 5 | 359 | |||
BC072656.1 | - | 16, 17 | 1658 | 30536315 |