Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phactr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161797.1 | NP_001155269.1 | 3 | 246 | ||
NM_175306.4 | NP_780515.2 | 3 | 246 | |||
GenBank mRNA | AK013935.1 | - | 1 | 1510 | ||
AK049209.1 | - | 3 | 209 | |||
AK220496.1 | - | 3 | 225 | |||
BB849287.1 | - | 3 | 239 | |||
BC075672.1 | - | 3 | 208 | 30549496 | ||
BC096033.1 | - | 3 | 165 | 6854767 | ||
BY294063.1 | - | 3 | 289 | |||
CN460852.1 | - | 3 | 198 |