Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pds5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175310.6 | NP_780519.3 | 2, 3 | 252 | ||
GenBank mRNA | AY102267.1 | - | 2, 3 | 252 | ||
BC096539.1 | - | 1, 2 | 115 | 6405163 | ||
BF471938.1 | - | 2, 3 | 155 |