Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spty2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175318.4 | NP_780527.2 | 2, 3 | 447 | ||
GenBank mRNA | AK030869.1 | - | 2, 3 | 282 | ||
AK147856.1 | - | 2, 3 | 256 | |||
AK162467.1 | - | 2, 3 | 262 | |||
BC079859.1 | - | 2, 3 | 242 | 6834544 |