Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175341.4 | NP_780550.1 | 2 | 822 | ||
NM_207515.2 | NP_997398.1 | 2 | 822 | |||
GenBank mRNA | AK028797.1 | - | 2 | 787 | ||
AK085709.1 | - | 2 | 785 | |||
AK148740.1 | - | 2 | 763 | |||
AK151913.1 | - | 2 | 786 | |||
AK151926.1 | - | 2 | 786 | |||
AK151957.1 | - | 2 | 786 | |||
AK153261.1 | - | 2 | 676 | |||
AK159213.1 | - | 2 | 780 | |||
AK164372.1 | - | 2 | 785 | |||
AK167479.1 | - | 2 | 652 | |||
AK167694.1 | - | 3 | 588 | |||
AK206292.1 | - | 1 | 64 | |||
AK216404.1 | - | 1 | 65 | |||
AK220470.1 | - | 796 | ||||
BC020406.1 | - | 1 | 220 | |||
BC023895.1 | - | 2 | 774 | |||
BC026711.1 | - | 1 | 187 | |||
BC075665.1 | - | 1 | 425 | 30605146 | ||
BC079898.1 | - | 2 | 771 | |||
GQ906534.1 | - | 1 | 140 |