Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc119b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175352.4 | NP_780561.1 | 1 | 145 | ||
GenBank mRNA | AK045128.1 | - | 1 | 96 | ||
AK082614.1 | - | 1 | 147 | |||
BC112388.1 | - | 1 | 117 | 5352914 |