Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tprn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175286.4 | NP_780495.2 | 1 | 1610 | ||
GenBank mRNA | AK082888.1 | - | 1 | 1591 | ||
AK165772.1 | - | 1 | 1552 | |||
AK171799.1 | - | 1 | 1624 | |||
BC055478.1 | - | 1 | 55 | |||
BC089508.1 | - | 1 | 1120 | 6589951 |