Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phactr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161797.1 | NP_001155269.1 | 3 | 297 | ||
NM_175306.4 | NP_780515.2 | 3 | 297 | |||
GenBank mRNA | AK049209.1 | - | 3 | 260 | ||
AK220496.1 | - | 3 | 276 | |||
BB849287.1 | - | 3 | 290 | |||
BC075672.1 | - | 3 | 259 | 30549496 | ||
CN460852.1 | - | 3 | 249 |