Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252530.1 | NP_001239459.1 | 14 | 2977 | ||
NM_001252531.1 | NP_001239460.1 | 14 | 2920 | |||
NM_175316.3 | NP_780525.1 | 14 | 2723 | |||
GenBank mRNA | AK033539.1 | - | 6 | 1349 | ||
AK048120.1 | - | 14 | 2724 | |||
AK087682.1 | - | 14 | 2975 | |||
AK132394.1 | - | 14 | 2978 | |||
AK163542.1 | - | 14 | 2973 | |||
AK173049.1 | - | 12 | 2509 |