Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175324.3 | NP_780533.2 | 11 | 1356 | ||
GenBank mRNA | AK077531.1 | - | 11 | 1357 | ||
AK128998.1 | - | 11 | 1335 | |||
AK139918.1 | - | 11 | 1300 | |||
AK145561.1 | - | 11 | 1345 | |||
AK145834.1 | - | 11 | 1336 | |||
BC006042.1 | - | 1 | 32 | |||
BC032994.1 | - | 4 | 285 | |||
BC045199.1 | - | 11 | 1320 | 5133197 |