Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsperd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175350.3 | NP_780559.2 | 18, 19 | 1680 | ||
GenBank mRNA | AK077082.1 | - | 9, 10 | 741 | ||
BC115864.1 | - | 7, 8 | 633 | |||
BC116633.1 | - | 8, 9 | 747 | 40056655 | ||
HQ441159.1 | - | 18, 19 | 1672 | |||
HQ441160.1 | - | 15, 16 | 1568 |