Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc119b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175352.4 | NP_780561.1 | 5 | 2151 | ||
GenBank mRNA | AK082614.1 | - | 5 | 2153 | ||
BC112388.1 | - | 5 | 2123 | 5352914 |