Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001216.2 | NP_001207.2 | 1 | 383 | ||
GenBank mRNA | AJ010588.1 | - | 1 | 350 | ||
AL554705.3 | - | 1 | 382 | |||
BC014950.1 | - | 1 | 445 | 4865275 | ||
EF122496.1 | - | 1 | 383 | |||
KU177952.1 | - | 1 | 341 | |||
KU177953.1 | - | 1 | 341 | |||
X66839.1 | - | 1 | 383 |