Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B630005N14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175312.4 | NP_780521.2 | 5 | 3240 | ||
GenBank mRNA | AK046745.1 | - | 5 | 3241 | ||
AK135664.1 | - | 1 | 605 | |||
AK164407.1 | - | 1 | 1460 |