Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252530.1 | NP_001239459.1 | 14 | 4495 | ||
NM_001252531.1 | NP_001239460.1 | 14 | 4438 | |||
NM_175316.3 | NP_780525.1 | 14 | 4241 | |||
GenBank mRNA | AK048120.1 | - | 14 | 4242 |