Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spty2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175318.4 | NP_780527.2 | 6 | 2443 | ||
GenBank mRNA | AK030869.1 | - | 6 | 2278 | ||
AK147856.1 | - | 6 | 2253 | |||
BC079859.1 | - | 6 | 2238 | 6834544 |