Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhlrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175340.4 | NP_780549.1 | 1 | 1808 | ||
GenBank mRNA | AK045746.1 | - | 1 | 1779 | ||
BC062644.1 | - | 1 | 1777 | |||
BC118951.1 | - | 1 | 1775 | 40093176 | ||
BC118952.1 | - | 1 | 1777 | 40093177 |