Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130674.2 | NP_001124146.1 | 7 | 1294 | ||
NM_001199671.1 | NP_001186600.1 | 8 | 2109 | |||
NM_001199672.1 | NP_001186601.1 | 8 | 2109 | |||
NM_001199673.1 | NP_001186602.1 | 6 | 1094 | |||
NM_001219.4 | NP_001210.1 | 7 | 1294 | |||
NR_074086.1 | 5 | 868 | ||||
XM_017012659.1 | XP_016868148.1 | 7 | 1659 | |||
GenBank mRNA | AF013759.1 | - | 7 | 1200 | ||
AK027460.1 | - | 7 | 1223 | |||
AK056338.1 | - | 8 | 1997 | |||
AK074862.1 | - | 7 | 1244 | |||
AK075089.1 | - | 3 | 561 | |||
BC013383.2 | - | 7 | 1193 | 3507961 | ||
HM002604.1 | - | 8 | 2110 | |||
HM002605.1 | - | 8 | 2110 | |||
HM002606.1 | - | 6 | 1036 | |||
HM002607.1 | - | 8 | 1144 | |||
HM002609.1 | - | 5 | 952 | |||
HM002610.1 | - | 5 | 811 | |||
HM002611.1 | - | 6 | 1094 | |||
HM002613.1 | - | 6 | 1099 | |||
HM002615.1 | - | 9 | 2304 | |||
HM002616.1 | - | 5 | 868 |