Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsperd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175350.3 | NP_780559.2 | 19 | 1719 | ||
GenBank mRNA | AK077082.1 | - | 10 | 780 | ||
BC115864.1 | - | 8 | 672 | |||
BC116633.1 | - | 9 | 786 | 40056655 | ||
HQ441159.1 | - | 19 | 1711 | |||
HQ441160.1 | - | 16 | 1607 |