Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam72a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175382.3 | NP_780591.1 | 1 | 813 | ||
GenBank mRNA | AK041641.1 | - | 1 | 815 | ||
AK076058.1 | - | 1 | 835 | |||
BC090647.1 | - | 1 | 899 | 30633261 | ||
BC106123.1 | - | 1 | 797 | 3995296 | ||
BC138225.1 | - | 1 | 782 | 8861244 | ||
BC145667.1 | - | 1 | 782 | 40130970 |