Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASQ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001232.3 | NP_001223.2 | 11 | 1678 | ||
GenBank mRNA | AK129831.1 | - | 3 | 533 | ||
AK129891.1 | - | 11 | 1653 | |||
BC022288.1 | - | 11 | 1587 | 4289141 | ||
D55655.1 | - | 11 | 1523 |