Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175446.3 | NP_780655.2 | 7 | 969 | ||
GenBank mRNA | AK039100.1 | - | 3 | 2075 | ||
AK046974.1 | - | 2 | 1101 | |||
AK133258.1 | - | 966 | ||||
AK220419.1 | - | 6 | 610 | |||
BC013473.1 | - | 4 | 322 | |||
BC067022.1 | - | 7 | 945 | |||
BC127247.1 | - | 2 | 538 | |||
BC128569.1 | - | 2 | 538 |