Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175484.3 | NP_780693.2 | 3 | 605 | ||
GenBank mRNA | AK047330.1 | - | 3 | 522 | ||
AK047527.1 | - | 3 | 547 | |||
AK087414.1 | - | 3 | 606 | |||
BB647296.1 | - | 3 | 542 | |||
BC062649.1 | - | 3 | 337 | 6416110 |