Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040059.1 | NP_001035148.1 | 6 | 1648 | ||
NM_001251.2 | NP_001242.2 | 6 | 1730 | |||
GenBank mRNA | BC015557.2 | - | 6 | 1602 | 3050731 | |
BX647524.1 | - | 6 | 1497 | |||
S57235.1 | - | 6 | 1560 |
Gene Symbol: LOC100996842 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI188961.1 | - | 1 | 15 | ||
R55342.1 | - | 1 | 28 |