Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081684.1 | NP_001075153.1 | 2 | 1985 | ||
NM_175428.3 | NP_780637.2 | 3 | 2128 | |||
GenBank mRNA | AK034057.1 | - | 3 | 2129 | ||
AK129313.1 | - | 2 | 1962 | |||
AK145072.1 | - | 2 | 1968 |